siRNA Details
Gene Symbol: Cdk5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109309.1 | NP_001102779.1 | 1 | 679 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009872, BC100715.
Gene Ontology Categories:
