siRNA Details
Gene Symbol: Gpr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245491.2 | XP_006245553.1 | 2 | 2839 | ||
XM_006245492.2 | XP_006245554.1 | 2 | 2457 | |||
XM_006245493.2 | XP_006245555.1 | 2 | 1644 | |||
XM_006245494.2 | XP_006245556.1 | 2 | 986 | |||
XM_006245495.2 | XP_006245557.1 | 2 | 2575 |
