siRNA Details
Gene Symbol: RGD1563692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109311.1 | NP_001102781.1 | 2 | 374 | ||
XM_006245547.2 | XP_006245609.1 | 1 | 1401 | |||
GenBank mRNA | BC162036.1 | - | 1 | 187 | 7097280 |
