siRNA Details
Gene Symbol: Sh2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109313.2 | NP_001102783.2 | 1, 2 | 363 | ||
XM_008773527.1 | XP_008771749.1 | 1, 2 | 502 | |||
XM_008773528.1 | XP_008771750.1 | 1, 2 | 493 | |||
GenBank mRNA | BC167036.1 | - | 1, 2 | 363 | 7389183 |
