siRNA Details
Gene Symbol: RGD1560927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109315.1 | NP_001102785.1 | 2 | 369 | ||
XM_006257482.1 | XP_006257544.1 | 2 | 274 | |||
GenBank mRNA | FQ221578.1 | - | 2 | 267 |
Gene Symbol: LOC103694509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773491.1 | XP_008771713.1 | 2 | 388 |
