siRNA Details
Gene Symbol: Fancb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192003.1 | NP_001178932.1 | 2 | 960 | ||
XM_006256884.2 | XP_006256946.1 | 3 | 1383 | |||
XM_006256885.2 | XP_006256947.1 | 4 | 1423 | |||
XM_006256886.2 | XP_006256948.1 | 3 | 1385 | |||
XM_008773184.1 | XP_008771406.1 | 3 | 1384 | |||
XM_008773185.1 | XP_008771407.1 | 5 | 1527 |
