siRNA Details
Gene Symbol: Slc10a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024368.1 | NP_001019539.1 | 3 | 1246 | ||
XM_006229606.2 | XP_006229668.1 | 3 | 2003 | |||
XM_008773611.1 | XP_008771833.1 | 3 | 1386 | |||
GenBank mRNA | BC086556.1 | - | 3 | 1246 | 7304843 |
