siRNA Details
Gene Symbol: Cd300a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205348.1 | NP_001192277.1 | 6 | 1072 | ||
GenBank mRNA | EF199812.1 | - | 6 | 767 | ||
FQ230743.1 | - | 6 | 837 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd300a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205348.1 | NP_001192277.1 | 6 | 1072 | ||
GenBank mRNA | EF199812.1 | - | 6 | 767 | ||
FQ230743.1 | - | 6 | 837 |