siRNA Details
Gene Symbol: Qrich2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247833.2 | XP_006247895.2 | 8 | 4563 | ||
XM_008768433.1 | XP_008766655.1 | 9 | 4533 | |||
XM_008768434.1 | XP_008766656.1 | 8 | 4233 | |||
XM_008768435.1 | XP_008766657.1 | 7 | 4023 | |||
XM_008768436.1 | XP_008766658.1 | 8 | 3963 | |||
XM_008768437.1 | XP_008766659.1 | 6 | 2792 |
