siRNA Details
Gene Symbol: Cd84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192006.1 | NP_001178935.1 | 5 | 849 | ||
XM_008769753.1 | XP_008767975.1 | 6 | 1264 | |||
XM_008769754.1 | XP_008767976.1 | 7 | 1331 | |||
XM_008769755.1 | XP_008767977.1 | 6 | 1242 | |||
XM_008769756.1 | XP_008767978.1 | 6 | 1241 | |||
XM_008769757.1 | XP_008767979.1 | 5 | 867 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013489, AB196814, AK171188, BC140952, AK171067, and others.
Gene Ontology Categories:
