siRNA Details
Gene Symbol: Cd84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192006.1 | NP_001178935.1 | 6 | 902 | ||
XM_008769753.1 | XP_008767975.1 | 7 | 1317 | |||
XM_008769754.1 | XP_008767976.1 | 8 | 1384 | |||
XM_008769755.1 | XP_008767977.1 | 7 | 1295 | |||
XM_008769756.1 | XP_008767978.1 | 7 | 1294 | |||
XM_008769757.1 | XP_008767979.1 | 6 | 920 |
