siRNA Details
Gene Symbol: RGD1563483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751309.2 | XP_003751357.1 | 3 | 642 |
Gene Symbol: LOC685907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770839.1 | XP_008769061.1 | 2 | 513 |
