siRNA Details
Gene Symbol: Slc2a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191551.1 | NP_001178480.1 | 4 | 400 | ||
XM_006251100.1 | XP_006251162.1 | 5 | 433 | |||
XM_006251101.2 | XP_006251163.1 | 5 | 539 | |||
XM_006251102.2 | XP_006251164.1 | 4 | 571 |
