siRNA Details
Gene Symbol: RGD1564308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253169.2 | XP_006253231.1 | 3 | 478 | ||
XM_006253170.2 | XP_006253232.1 | 3 | 263 | |||
XM_577523.6 | XP_577523.4 | 3 | 494 |
