siRNA Details
Gene Symbol: Prl2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044271.1 | NP_001037736.1 | 5 | 696 | ||
GenBank mRNA | DQ329281.1 | - | 5 | 696 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prl2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044271.1 | NP_001037736.1 | 5 | 696 | ||
GenBank mRNA | DQ329281.1 | - | 5 | 696 |