siRNA Details
Gene Symbol: Prl2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044271.1 | NP_001037736.1 | 1, 2 | 93 | ||
GenBank mRNA | DQ329281.1 | - | 1, 2 | 93 |

More Information
Other Species Targeted

Gene Ontology Categories:
