siRNA Details
Gene Symbol: Lrrc29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195616.1 | NP_001182545.1 | 3 | 583 | ||
XM_006255531.2 | XP_006255593.1 | 3 | 818 | |||
XM_006255532.2 | XP_006255594.1 | 3 | 726 | |||
XM_006255533.2 | XP_006255595.1 | 3 | 662 | |||
GenBank mRNA | BC166813.1 | - | 3 | 583 | 7454853 |
