siRNA Details
Gene Symbol: Tbc1d22b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025059.1 | NP_001020230.1 | 3 | 216 | ||
GenBank mRNA | AY380821.1 | - | 3 | 216 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tbc1d22b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025059.1 | NP_001020230.1 | 3 | 216 | ||
GenBank mRNA | AY380821.1 | - | 3 | 216 |