siRNA Details
Gene Symbol: RGD1563348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114939.1 | NP_001108411.1 | 1 | 120 | ||
GenBank mRNA | BI285703.1 | - | 4 | 552 | ||
FQ220990.1 | - | 1 | 129 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1563348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114939.1 | NP_001108411.1 | 1 | 120 | ||
GenBank mRNA | BI285703.1 | - | 4 | 552 | ||
FQ220990.1 | - | 1 | 129 |