siRNA Details
Gene Symbol: RGD1563348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114939.1 | NP_001108411.1 | 1, 2 | 195 | ||
GenBank mRNA | AI030980.1 | - | 3, 4 | 455 | ||
BI285703.1 | - | 3, 4 | 477 | |||
FQ220990.1 | - | 1, 2 | 204 |
