siRNA Details
Gene Symbol: Lrrc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025155.2 | NP_001020326.1 | 2 | 515 | ||
XM_008762657.1 | XP_008760879.1 | 2 | 803 | |||
GenBank mRNA | BC079098.1 | - | 2 | 392 | 7124390 | |
DY314273.1 | - | 2 | 515 |
