siRNA Details
Gene Symbol: Vwde | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762754.1 | XP_008760976.1 | 17 | 3335 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vwde | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762754.1 | XP_008760976.1 | 17 | 3335 |