siRNA Details
Gene Symbol: Agk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127497.1 | NP_001120969.1 | 10, 11 | 731 | ||
XM_006236363.2 | XP_006236425.1 | 10, 11 | 1033 | |||
XM_008762821.1 | XP_008761043.1 | 10, 11 | 1128 |
