siRNA Details
Gene Symbol: Tas2r143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025061.1 | NP_001020232.1 | 1 | 794 |
Gene Symbol: LOC103690198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762927.1 | XP_008761149.1 | 1 | 1122 |
