siRNA Details
Gene Symbol: Scrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025063.1 | NP_001020234.1 | 2 | 231 | ||
XM_006236512.2 | XP_006236574.1 | 2 | 219 | |||
XM_008762916.1 | XP_008761138.1 | 2 | 83 | |||
GenBank mRNA | BC079152.1 | - | 2 | 231 | 7128646 |
