siRNA Details
Gene Symbol: Jagn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044272.1 | NP_001037737.1 | 2 | 233 | ||
XM_006237053.2 | XP_006237115.1 | 2 | 282 | |||
GenBank mRNA | BC098749.1 | - | 2 | 233 | 7365215 | |
FQ225993.1 | - | 2 | 237 |
