siRNA Details
Gene Symbol: Ctnnbip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173388.1 | NP_001166859.1 | 4 | 350 | ||
XM_006239436.2 | XP_006239498.1 | 3 | 337 | |||
GenBank mRNA | BC098773.1 | - | 3 | 200 | ||
CB611569.1 | - | 4 | 367 |
