siRNA Details
Gene Symbol: RGD1565410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113792.1 | NP_001107264.1 | 369 | |||
XM_006241770.2 | XP_006241832.1 | 4 | 387 | |||
GenBank mRNA | BC158674.1 | - | 369 | 8371673 |
