siRNA Details
Gene Symbol: Glb1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192018.1 | NP_001178947.1 | 3, 4 | 333 | ||
XM_006242738.2 | XP_006242800.1 | 2, 3 | 371 | |||
XM_008766062.1 | XP_008764284.1 | 4, 5 | 1463 | |||
XM_008766063.1 | XP_008764285.1 | 3, 4 | 347 | |||
XM_008766064.1 | XP_008764286.1 | 2, 3 | 443 |
