siRNA Details
Gene Symbol: Rbpms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173426.1 | NP_001166897.1 | 3 | 281 | ||
XM_006243274.2 | XP_006243336.1 | 3 | 1967 | |||
XM_008766329.1 | XP_008764551.1 | 2 | 390 | |||
GenBank mRNA | BC169038.1 | - | 3 | 281 | 9090232 |
