siRNA Details
Gene Symbol: Lyg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098838.1 | NP_001092308.1 | 234 |
Gene Symbol: LOC100910070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244868.1 | XP_006244930.1 | 3 | 176 |
