siRNA Details
Gene Symbol: Tpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019353.2 | NP_062226.2 | 15 | 2601 | ||
XM_008764607.1 | XP_008762829.1 | 14 | 2559 | |||
XM_008764608.1 | XP_008762830.1 | 15 | 2595 | |||
GenBank mRNA | M31655.1 | - | 11 | 2127 | ||
X17396.1 | - | 15 | 2600 |
