siRNA Details
Gene Symbol: Enpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019370.2 | NP_062243.2 | 18, 19 | 1797 | ||
GenBank mRNA | BC097326.1 | - | 18, 19 | 1797 | 7458431 | |
D30649.1 | - | 18, 19 | 1824 | |||
FQ218696.1 | - | 1, 2 | 182 | |||
U78787.1 | - | 18, 19 | 1794 | |||
U78788.1 | - | 18, 19 | 1800 | |||
Z47987.1 | - | 18, 19 | 1781 |
