siRNA Details
Gene Symbol: Gpr34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024925.2 | NP_001020096.1 | 3 | 960 | ||
XM_006256675.2 | XP_006256737.1 | 4 | 3593 | |||
XM_006256676.2 | XP_006256738.1 | 4 | 1510 | |||
GenBank mRNA | AB159443.1 | - | 3 | 960 |
