siRNA Details
Gene Symbol: Srgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020074.2 | NP_064459.1 | 2 | 130 | ||
XM_006256483.2 | XP_006256545.1 | 5 | 510 | |||
XM_006256484.2 | XP_006256546.1 | 3 | 318 | |||
GenBank mRNA | BC088144.1 | - | 2 | 130 | 7384101 | |
FQ220127.1 | - | 2 | 149 | |||
FQ227535.1 | - | 2 | 147 | |||
FQ231972.1 | - | 2 | 147 | |||
J03224.1 | - | 2 | 143 | |||
K02934.1 | - | 2 | 145 |
