siRNA Details
Gene Symbol: Haao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020076.2 | NP_064461.1 | 7 | 675 | ||
XM_006239684.2 | XP_006239746.1 | 7 | 1580 | |||
GenBank mRNA | BC085739.1 | - | 7 | 629 | 7108551 | |
D28339.1 | - | 5 | 389 | |||
D44494.1 | - | 7 | 676 | |||
FQ219702.1 | - | 7 | 689 |
