siRNA Details
Gene Symbol: Adam17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020306.2 | NP_064702.1 | 5 | 815 | ||
GenBank mRNA | AJ012603.1 | - | 5 | 817 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adam17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020306.2 | NP_064702.1 | 5 | 815 | ||
GenBank mRNA | AJ012603.1 | - | 5 | 817 |