siRNA Details
Gene Symbol: Lair1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029928.1 | NP_001025099.1 | 5, 6 | 593 | ||
XM_008758974.1 | XP_008757196.1 | 7, 8 | 1086 | |||
GenBank mRNA | AY863023.1 | - | 5, 6 | 593 |
