siRNA Details
Gene Symbol: Ptges | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021583.3 | NP_067594.1 | 3 | 337 | ||
GenBank mRNA | AB035145.1 | - | 3 | 297 | ||
AB041998.1 | - | 3 | 326 | |||
AB048730.1 | - | 3 | 339 | |||
AF280967.1 | - | 3 | 336 | |||
BC088101.1 | - | 3 | 295 | |||
BC105858.1 | - | 3 | 323 | 7456259 |
