siRNA Details
Gene Symbol: LOC606294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031627.1 | NP_001026797.1 | 7 | 1441 | ||
GenBank mRNA | BC060521.1 | - | 6 | 1334 | ||
BC098692.1 | - | 7 | 1441 | 7386167 |
