siRNA Details
Gene Symbol: Max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022210.1 | NP_071546.1 | 3 | 119 | ||
XM_006240248.2 | XP_006240310.1 | 2 | 318 | |||
XM_008764846.1 | XP_008763068.1 | 3 | 350 | |||
GenBank mRNA | BC099778.1 | - | 3 | 281 | 7933028 | |
D14447.1 | - | 3 | 119 | |||
D14448.1 | - | 2 | 92 |
