siRNA Details
Gene Symbol: Max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022210.1 | NP_071546.1 | 1, 2 | 43 | ||
XM_008764846.1 | XP_008763068.1 | 1, 2 | 274 | |||
GenBank mRNA | BC099778.1 | - | 1, 2 | 205 | 7933028 | |
D14447.1 | - | 1, 2 | 43 |
