siRNA Details
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034995.1 | NP_001030167.1 | 7 | 1356 | ||
XM_006254636.2 | XP_006254698.1 | 7 | 1450 | |||
XM_006254637.2 | XP_006254699.1 | 7 | 1764 | |||
XM_006254639.2 | XP_006254701.1 | 8 | 1561 | |||
XM_006254640.2 | XP_006254702.1 | 6 | 1300 | |||
XM_006254641.2 | XP_006254703.1 | 5 | 966 | |||
XM_008772069.1 | XP_008770291.1 | 7 | 1316 | |||
GenBank mRNA | BC103648.1 | - | 7 | 1356 | 7315158 | |
BC166497.1 | - | 4 | 827 |
