siRNA Details
Gene Symbol: Ctps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034998.2 | NP_001030170.1 | 12 | 1322 | ||
XM_006256895.2 | XP_006256957.1 | 12 | 1324 | |||
XM_006256896.2 | XP_006256958.1 | 12 | 1612 | |||
XM_006256897.2 | XP_006256959.1 | 12 | 1445 | |||
XM_006256898.2 | XP_006256960.1 | 12 | 1406 | |||
XM_006256899.2 | XP_006256961.1 | 12 | 1777 | |||
XM_006256900.2 | XP_006256962.1 | 12 | 1420 | |||
GenBank mRNA | BC085358.1 | - | 5 | 424 | ||
BC085949.1 | - | 12 | 1322 | 7132135 |
