siRNA Details
Gene Symbol: Nicn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034999.1 | NP_001030171.1 | 2 | 224 | ||
XM_006243854.2 | XP_006243916.1 | 2 | 308 | |||
GenBank mRNA | BC101861.1 | - | 2 | 224 | 7936966 | |
FQ229605.1 | - | 2 | 113 |
