siRNA Details
Gene Symbol: Ctgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022266.2 | NP_071602.1 | 5 | 1017 | ||
GenBank mRNA | AB023068.1 | - | 5 | 1005 | ||
AF079531.1 | - | 2 | 151 | |||
AF120275.1 | - | 5 | 1017 | |||
AY596447.1 | - | 1 | 1578 | |||
BC072503.1 | - | 4 | 1386 | 7098774 |
