siRNA Details
Gene Symbol: Ctgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022266.2 | NP_071602.1 | 2 | 454 | ||
GenBank mRNA | AB023068.1 | - | 2 | 442 | ||
AF120275.1 | - | 2 | 454 | |||
AY596447.1 | - | 1 | 321 | |||
BC072503.1 | - | 2 | 444 | 7098774 |
