siRNA Details
Gene Symbol: LOC641544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047113.1 | NP_001040578.1 | 664 | |||
GenBank mRNA | AY240937.2 | - | 664 |

More Information
Other Species Targeted

