siRNA Details
Gene Symbol: Sptan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171983.2 | NP_741984.2 | 45 | 5964 | ||
XM_008761673.1 | XP_008759895.1 | 46 | 5979 | |||
XM_008761674.1 | XP_008759896.1 | 45 | 5964 | |||
XM_008761675.1 | XP_008759897.1 | 45 | 5919 | |||
XM_008761676.1 | XP_008759898.1 | 46 | 5979 | |||
XM_008761677.1 | XP_008759899.1 | 45 | 5919 | |||
XM_008761678.1 | XP_008759900.1 | 44 | 5904 | |||
GenBank mRNA | AF084186.1 | - | 44 | 5856 | ||
BC070885.1 | - | 44 | 5868 | 7098134 | ||
GQ502182.2 | - | 45 | 5871 | |||
J04828.1 | - | 15 | 1983 | |||
M19726.1 | - | 6 | 547 | |||
X90845.1 | - | 45 | 5964 |
