siRNA Details
Gene Symbol: Fxyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022388.1 | NP_071783.1 | 6, 7 | 1128 | ||
XM_006237206.2 | XP_006237268.1 | 7, 8 | 467 | |||
XM_008763260.1 | XP_008761482.1 | 7, 8 | 797 | |||
XM_008763261.1 | XP_008761483.1 | 7, 8 | 755 | |||
GenBank mRNA | L41254.1 | - | 6, 7 | 1128 |
